siRNA Details
| Gene Symbol: Diap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172493.2 | NP_766081.1 | 1 | 359 | ||
| GenBank mRNA | AB713187.1 | - | 1 | 133 | ||
| AK088648.1 | - | 1 | 305 | |||
| AK153986.1 | - | 1 | 359 | |||
| AY312280.1 | - | 1 | 296 | |||
| BC100759.1 | - | 1 | 277 | |||

Diap2
Genomic Map