siRNA Details
| Gene Symbol: Snx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017472.4 | NP_059500.2 | 1, 2 | 440 | ||
| GenBank mRNA | AF062482.1 | - | 1, 2 | 238 | ||
| AK007798.1 | - | 1, 2 | 251 | |||
| AK013183.1 | - | 1, 2 | 465 | |||
| AK013261.1 | - | 1, 2 | 73 | |||
| AK018790.1 | - | 1, 2 | 73 | |||
| AK145350.1 | - | 1, 2 | 243 | |||
| AK154728.1 | - | 1, 2 | 243 | |||
| AK189751.1 | - | 1, 2 | 31 | |||
| BC116885.1 | - | 1, 2 | 284 | 40126204 | ||
| BC116889.1 | - | 1, 2 | 284 | 40126208 | ||

Snx3
Genomic Map