siRNA Details
| Gene Symbol: Rp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018739.2 | NP_061209.1 | 3, 4 | 436 | ||
| GenBank mRNA | AK146136.1 | - | 3, 4 | 417 | ||
| AK147912.1 | - | 439 | ||||
| AK168391.1 | - | 3, 4 | 429 | |||
| BC048480.1 | - | 3, 4 | 294 | |||
| BC115549.1 | - | 3, 4 | 396 | 40101700 | ||
| BC115550.1 | - | 3, 4 | 396 | 40101701 | ||
| BY239981.1 | - | 3 | 437 | |||
| D78255.1 | - | 3, 4 | 415 | |||

Rp9
Genomic Map