siRNA Details
| Gene Symbol: Prkx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016979.1 | NP_058675.1 | 9 | 1825 | ||
| GenBank mRNA | AK034681.1 | - | 9 | 2144 | ||
| AK037141.1 | - | 9 | 1820 | |||
| AK086026.1 | - | 2 | 507 | |||
| AK139510.1 | - | 9 | 1833 | |||
| AK149937.1 | - | 9 | 1803 | |||
| AK150588.1 | - | 9 | 1787 | |||
| AK152166.1 | - | 9 | 1621 | |||
| AK154447.1 | - | 9 | 1805 | |||
| AK169322.1 | - | 9 | 1776 | |||
| BC006875.1 | - | 9 | 1825 | 3596013 | ||

Prkx
Genomic Map