siRNA Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289492.1 | NP_001276421.1 | 7 | 1170 | ||
| NM_001289493.1 | NP_001276422.1 | 6 | 995 | |||
| NM_018734.3 | NP_061204.3 | 6 | 1032 | |||
| GenBank mRNA | AK132266.1 | - | 6 | 1031 | ||
| BC019195.1 | - | 7 | 1081 | 5036920 | ||
| U44731.1 | - | 6 | 884 | |||

Gbp3
Genomic Map