siRNA Details
| Gene Symbol: Golga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018748.3 | NP_061218.2 | 2 | 277 | ||
| GenBank mRNA | AF051357.2 | - | 2 | 276 | ||
| AK161568.1 | - | 2 | 329 | |||
| AK172292.1 | - | 2 | 325 | |||
| BC003268.1 | - | 2 | 277 | |||
| BC023102.1 | - | 2 | 212 | |||
| BC037641.1 | - | 2 | 322 | |||
| BC057561.1 | - | 2 | 144 | |||
| BC096467.1 | - | 2 | 277 | |||

Golga4
Genomic Map