siRNA Details
| Gene Symbol: Sgsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018822.3 | NP_061292.2 | 2 | 134 | ||
| GenBank mRNA | AF153827.1 | - | 2 | 108 | ||
| AF304053.1 | - | 2 | 108 | |||
| AF304054.1 | - | 2 | 108 | |||
| AK052266.1 | - | 2 | 127 | |||
| AK076267.1 | - | 2 | 163 | |||
| AK128926.1 | - | 2 | 134 | |||
| AK141266.1 | - | 2 | 82 | |||
| BC130268.1 | - | 2 | 131 | 40143074 | ||

Sgsh
Genomic Map