siRNA Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018830.1 | NP_061300.1 | 6 | 581 | ||
| GenBank mRNA | AB037111.1 | - | 7 | 866 | ||
| AB037181.1 | - | 6 | 581 | |||
| AK034493.1 | - | 6 | 812 | |||
| AK041937.1 | - | 6 | 770 | |||
| AK046540.1 | - | 6 | 775 | |||
| AK047692.1 | - | 6 | 769 | |||
| AK080951.1 | - | 6 | 758 | |||
| AK166100.1 | - | 6 | 829 | |||
| BC137898.1 | - | 6 | 564 | 8860917 | ||
| BC145016.1 | - | 6 | 567 | 9053564 | ||

Asah2
Genomic Map