siRNA Details
| Gene Symbol: Magix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018832.2 | NP_061302.2 | 2, 3 | 294 | ||
| GenBank mRNA | AF229645.1 | - | 2, 3 | 314 | ||
| AK077898.1 | - | 2, 3 | 261 | |||
| AK158091.1 | - | 2, 3 | 234 | |||
| BC099459.1 | - | 2, 3 | 294 | 30847792 | ||
| BC132580.1 | - | 2, 3 | 233 | 40130857 | ||
| BC132582.1 | - | 2, 3 | 233 | 40130859 | ||

Magix
Genomic Map