siRNA Details
| Gene Symbol: Tcstv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018756.3 | NP_061226.1 | 2 | 305 | ||
| GenBank mRNA | AF067057.1 | - | 2 | 287 | ||
| AF067059.1 | - | 2 | 305 | |||
| BC119476.2 | - | 1 | 789 | 40049850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcstv1| Gene Symbol: Tcstv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018756.3 | NP_061226.1 | 2 | 305 | ||
| GenBank mRNA | AF067057.1 | - | 2 | 287 | ||
| AF067059.1 | - | 2 | 305 | |||
| BC119476.2 | - | 1 | 789 | 40049850 | ||