siRNA Details
| Gene Symbol: Cd160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163496.1 | NP_001156968.1 | 3, 4 | 609 | ||
| NM_018767.3 | NP_061237.3 | 3, 4 | 532 | |||
| GenBank mRNA | AF060982.1 | - | 3, 4 | 503 | ||
| AK042093.1 | - | 3, 4 | 533 | |||
| AK139789.1 | - | 3, 4 | 610 | |||
| AK145332.1 | - | 3, 4 | 590 | |||
| BC021596.1 | - | 3, 4 | 504 | 5098004 | ||

Cd160
Genomic Map