siRNA Details
| Gene Symbol: Cldn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018777.4 | NP_061247.1 | 3 | 704 | ||
| GenBank mRNA | AF087824.1 | - | 1 | 581 | ||
| AF125305.1 | - | 1 | 581 | |||
| AK010560.1 | - | 3 | 705 | |||
| AK010682.1 | - | 3 | 699 | |||
| BC005718.1 | - | 3 | 681 | 3594823 | ||
| BC050138.1 | - | 3 | 687 | 30028814 | ||
| DQ864649.1 | - | 1 | 581 | |||

Cldn6
Genomic Map