siRNA Details
| Gene Symbol: Foxo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018789.2 | NP_061259.1 | 1, 2 | 746 | ||
| GenBank mRNA | AB032770.1 | - | 1, 2 | 526 | ||
| AB221664.1 | - | 1, 2 | 453 | |||
| AF114260.1 | - | 1, 2 | 746 | |||
| BC139275.1 | - | 1, 2 | 587 | 8862297 | ||
| BC139304.1 | - | 1, 2 | 587 | 8862326 | ||

Foxo4
Genomic Map