siRNA Details
| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018810.2 | NP_061280.2 | 2 | 308 | ||
| GenBank mRNA | AB026986.1 | - | 2 | 413 | ||
| AF192785.1 | - | 2 | 333 | |||
| AK005137.1 | - | 2 | 142 | |||
| AK029500.1 | - | 2 | 348 | |||
| AK077060.1 | - | 2 | 309 | |||
| AK078962.1 | - | 2 | 238 | |||
| BC003329.1 | - | 2 | 297 | 3158135 | ||

Mkrn1
Genomic Map