siRNA Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018811.6 | NP_061281.3 | 6 | 860 | ||
| GenBank mRNA | AF201730.1 | - | 6 | 849 | ||
| AF546701.1 | - | 6 | 861 | |||
| AK008690.1 | - | 6 | 870 | |||
| AK016296.1 | - | 4 | 521 | |||
| AK139526.1 | - | 6 | 869 | |||
| AK153642.1 | - | 6 | 907 | |||
| BC027098.1 | - | 6 | 830 | 5356667 | ||
| BC089477.1 | - | 6 | 840 | 6741521 | ||

Abhd2
Genomic Map