siRNA Details
| Gene Symbol: Irx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018826.2 | NP_061296.1 | 2 | 960 | ||
| GenBank mRNA | AF165985.1 | - | 2 | 572 | ||
| AF230074.1 | - | 2 | 1127 | |||
| AJ271054.1 | - | 2 | 353 | |||
| AK004747.1 | - | 2 | 773 | |||
| BC051959.1 | - | 2 | 753 | 6404946 | ||
| BC056994.1 | - | 2 | 960 | 5716910 | ||

Irx5
Genomic Map