siRNA Details
| Gene Symbol: Golga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018748.3 | NP_061218.2 | 15 | 3152 | ||
| GenBank mRNA | AF051357.2 | - | 15 | 3151 | ||
| BC053000.1 | - | 1 | 16 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Golga4| Gene Symbol: Golga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018748.3 | NP_061218.2 | 15 | 3152 | ||
| GenBank mRNA | AF051357.2 | - | 15 | 3151 | ||
| BC053000.1 | - | 1 | 16 | |||