siRNA Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018749.2 | NP_061219.2 | 14, 15 | 1795 | ||
| GenBank mRNA | AB012580.1 | - | 14, 15 | 1736 | ||
| AK045228.1 | - | 14, 15 | 1737 | |||
| AK049767.1 | - | 14, 15 | 1734 | |||
| AK145689.1 | - | 14, 15 | 1750 | |||
| AK168256.1 | - | 14, 15 | 1785 | |||
| AK191549.1 | - | 1, 2 | 92 | |||
| BC018598.1 | - | 14, 15 | 1733 | |||
| BC089020.1 | - | 14, 15 | 1729 | 30551991 | ||

Eif3d
Genomic Map