siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018759.2 | NP_061229.2 | 11 | 1437 | ||
| GenBank mRNA | AB012725.1 | - | 11 | 1403 | ||
| AK017693.1 | - | 11 | 1438 | |||
| AK162729.1 | - | 5 | 441 | |||
| BC037055.1 | - | 9 | 978 | 4948371 | ||
| BC039041.1 | - | 10 | 3442 | |||

Zfp326
Genomic Map