siRNA Details
| Gene Symbol: Tbc1d8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018775.4 | NP_061245.3 | 19 | 3102 | ||
| GenBank mRNA | AK138630.1 | - | 19 | 3100 | ||
| AK141520.1 | - | 19 | 3091 | |||
| AK141816.1 | - | 19 | 3057 | |||
| AK162371.1 | - | 13 | 1877 | |||
| BC005421.1 | - | 16 | 2437 | |||
| BC065081.1 | - | 19 | 3036 | 6832495 | ||
| U88873.1 | - | 15 | 2322 | |||

Tbc1d8
Genomic Map