siRNA Details
| Gene Symbol: KCNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112741.1 | NP_001106212.1 | 2 | 873 | ||
| NM_004976.4 | NP_004967.1 | 2 | 873 | |||
| XM_011520080.2 | XP_011518382.1 | 2 | 2011 | |||
| GenBank mRNA | BC107129.2 | - | 2 | 890 | 40024733 | |
| L00621.1 | - | 2 | 886 | |||
| M96747.1 | - | 1 | 89 | |||
| S56770.1 | - | 2 | 886 | |||

KCNC1
Genomic Map