siRNA Details
| Gene Symbol: Dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB028272.1 | - | 3 | 1543 | ||
| AK135947.1 | - | 2 | 949 | |||
| AK146657.1 | - | 3 | 1506 | |||
| AK158530.1 | - | 3 | 1403 | |||
| AK160924.1 | - | 3 | 1440 | |||
| AK167718.1 | - | 3 | 1443 | |||
| AK168220.1 | - | 3 | 1457 | |||
| AK168291.1 | - | 3 | 1457 | |||
| BC012962.1 | - | 3 | 1442 | 4188579 | ||

Dnajb1
Genomic Map