siRNA Details
| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018810.2 | NP_061280.2 | 8 | 2510 | ||
| GenBank mRNA | AF192785.1 | - | 8 | 2536 | ||
| AK029500.1 | - | 8 | 2550 | |||
| AK077060.1 | - | 8 | 2511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mkrn1| Gene Symbol: Mkrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018810.2 | NP_061280.2 | 8 | 2510 | ||
| GenBank mRNA | AF192785.1 | - | 8 | 2536 | ||
| AK029500.1 | - | 8 | 2550 | |||
| AK077060.1 | - | 8 | 2511 | |||