siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 5, 6 | 722 | ||
| GenBank mRNA | AF113751.1 | - | 5, 6 | 690 | ||
| AK053417.1 | - | 5, 6 | 723 | |||
| AK157272.1 | - | 5, 6 | 714 | |||
| AK173056.1 | - | 5, 6 | 676 | |||
| BC052468.1 | - | 5, 6 | 715 | 6835038 | ||

Nup210
Genomic Map