siRNA Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207678.1 | NP_997561.1 | 11 | 1712 | ||
| GenBank mRNA | AB041605.1 | - | 11 | 1689 | ||
| AK008585.1 | - | 6 | 706 | |||
| AK048244.1 | - | 11 | 3492 | |||
| AK193127.1 | - | 1 | 197 | |||
| BC003773.1 | - | 4 | 835 | |||
| BC023747.1 | - | 11 | 3480 | |||
| BC083055.1 | - | 4 | 699 | |||
| BC132295.1 | - | 11 | 1704 | 40130572 | ||
| U37351.1 | - | 4 | 718 | |||

Ccnl2
Genomic Map