siRNA Details
| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 2, 3 | 422 | ||
| GenBank mRNA | AF060565.1 | - | 2, 3 | 124 | ||
| AK139317.1 | - | 2, 3 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem131| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 2, 3 | 422 | ||
| GenBank mRNA | AF060565.1 | - | 2, 3 | 124 | ||
| AK139317.1 | - | 2, 3 | 423 | |||