siRNA Details
| Gene Symbol: Adgrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198894.1 | NP_001185823.1 | 3 | 514 | ||
| NM_018882.3 | NP_061370.2 | 3 | 464 | |||
| GenBank mRNA | AF166382.1 | - | 3 | 514 | ||
| AK087268.1 | - | 3 | 266 | |||
| AK133678.1 | - | 3 | 231 | |||
| AK141886.1 | - | 3 | 493 | |||
| AK141894.1 | - | 3 | 277 | |||
| AK164776.1 | - | 3 | 288 | |||
| AK167547.1 | - | 2 | 218 | |||
| BC034678.1 | - | 3 | 260 | 3709247 | ||
More Information
Set Membership: |
|

Adgrg1
Genomic Map