siRNA Details
Gene Symbol: Olfr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019473.1 | NP_062346.1 | 1 | 1031 | ||
GenBank mRNA | BC130252.1 | - | 1 | 1035 | 40142475 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019473.1 | NP_062346.1 | 1 | 1031 | ||
GenBank mRNA | BC130252.1 | - | 1 | 1035 | 40142475 |