siRNA Details
| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040699.1 | NP_001035789.1 | 6 | 733 | ||
| GenBank mRNA | AK028160.1 | - | 6 | 699 | ||
| AK081973.1 | - | 6 | 719 | |||
| AK148768.1 | - | 6 | 734 | |||
| BC032254.1 | - | 6 | 735 | 5368816 | ||

Mtmr7
Genomic Map