siRNA Details
| Gene Symbol: Ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019443.2 | NP_062316.1 | 2, 3 | 348 | ||
| GenBank mRNA | AK007840.1 | - | 2, 3 | 255 | ||
| AK131584.1 | - | 547 | ||||
| BC024364.1 | - | 2, 3 | 257 | 5031206 | ||
| CF383819.1 | - | 1, 2 | 132 | |||
| CT010398.1 | - | 2, 3 | 200 | |||
| Y07708.1 | - | 2, 3 | 367 | |||

Ndufa1
Genomic Map