siRNA Details
| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 11, 12 | 1243 | ||
| GenBank mRNA | AB093221.1 | - | 7, 8 | 679 | ||
| AF060565.1 | - | 11, 12 | 945 | |||
| AK139317.1 | - | 11, 12 | 1245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem131| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018872.2 | NP_061360.2 | 11, 12 | 1243 | ||
| GenBank mRNA | AB093221.1 | - | 7, 8 | 679 | ||
| AF060565.1 | - | 11, 12 | 945 | |||
| AK139317.1 | - | 11, 12 | 1245 | |||