siRNA Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018878.3 | NP_061366.2 | 6 | 997 | ||
| GenBank mRNA | AF104261.1 | - | 6 | 997 | ||
| AK144606.1 | - | 6 | 859 | |||
| AK148141.1 | - | 6 | 866 | |||
| AK205117.1 | - | 1 | 106 | |||
| AK214131.1 | - | 1 | 195 | |||
| BC066014.1 | - | 6 | 786 | 5704566 | ||

Paxip1
Genomic Map