siRNA Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018885.2 | NP_061373.1 | 6 | 2558 | ||
| GenBank mRNA | AF124732.1 | - | 6 | 2262 | ||
| AK132571.1 | - | 6 | 2558 | |||
| BB675268.1 | - | 1 | 364 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irx4| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018885.2 | NP_061373.1 | 6 | 2558 | ||
| GenBank mRNA | AF124732.1 | - | 6 | 2262 | ||
| AK132571.1 | - | 6 | 2558 | |||
| BB675268.1 | - | 1 | 364 | |||