siRNA Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019426.2 | NP_062299.2 | 2 | 1433 | ||
| GenBank mRNA | AJ132702.1 | - | 2 | 1426 | ||
| AK132359.1 | - | 1 | 53 | |||
| AK134252.1 | - | 2 | 1440 | |||
| AK167542.1 | - | 3 | 1479 | |||
| BC022714.1 | - | 2 | 1426 | |||
| BC037617.1 | - | 1435 | ||||
| BC048827.1 | - | 3 | 1594 | |||
| BC052460.1 | - | 2 | 1433 | 6834550 | ||
| BC060246.1 | - | 2 | 1416 | 5705945 | ||

Atf7ip
Genomic Map