siRNA Details
| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040699.1 | NP_001035789.1 | 13, 14 | 1701 | ||
| GenBank mRNA | AF073882.1 | - | 6, 7 | 760 | ||
| AK028160.1 | - | 13, 14 | 1667 | |||
| AK081973.1 | - | 13, 14 | 1687 | |||
| BC032254.1 | - | 14, 15 | 1995 | 5368816 | ||
| BC038468.1 | - | 3, 4 | 368 | |||

Mtmr7
Genomic Map