siRNA Details
| Gene Symbol: Unc93b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161428.1 | NP_001154900.1 | 7 | 875 | ||
| NM_019449.2 | NP_062322.2 | 7 | 875 | |||
| GenBank mRNA | AJ422144.1 | - | 1 | 17 | ||
| AK030488.1 | - | 7 | 836 | |||
| AK155556.1 | - | 7 | 833 | |||
| AK162535.1 | - | 7 | 828 | |||
| AK165060.1 | - | 7 | 835 | |||
| BC018388.1 | - | 7 | 842 | 4209296 | ||
| BC023731.1 | - | 1 | 300 | 5352340 | ||

Unc93b1
Genomic Map