siRNA Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019454.3 | NP_062327.2 | 7 | 1286 | ||
| GenBank mRNA | AB043893.1 | - | 7 | 929 | ||
| AF253469.1 | - | 7 | 1241 | |||
| AF273454.1 | - | 7 | 1307 | |||
| AK004739.1 | - | 7 | 1254 | |||
| AK031699.1 | - | 7 | 1255 | |||
| AK052322.1 | - | 7 | 1287 | |||
| AK165517.1 | - | 7 | 1254 | |||
| AK171717.1 | - | 7 | 1253 | |||
| BC042497.1 | - | 7 | 1241 | 4017786 | ||
| BC049130.1 | - | 7 | 1246 | 6825525 | ||

Dll4
Genomic Map