siRNA Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081549.2 | NP_001075018.1 | 4 | 902 | ||
| NM_019466.4 | NP_062339.2 | 4 | 748 | |||
| GenBank mRNA | AF263239.1 | - | 4 | 682 | ||
| AF263240.1 | - | 4 | 765 | |||
| AK010696.1 | - | 619 | ||||
| AK077283.1 | - | 4 | 748 | |||
| AK146764.1 | - | 4 | 877 | |||
| AY325904.1 | - | 4 | 858 | |||
| BC013551.1 | - | 4 | 728 | 4236038 | ||

Rcan1
Genomic Map