siRNA Details
| Gene Symbol: Ptbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019550.2 | NP_062423.1 | 4 | 332 | ||
| GenBank mRNA | AF095718.2 | - | 4 | 202 | ||
| AK041655.1 | - | 4 | 231 | |||
| AK050624.1 | - | 4 | 213 | |||
| AK050650.1 | - | 4 | 213 | |||
| AK135738.1 | - | 4 | 333 | |||
| AK137348.1 | - | 4 | 211 | |||
| AK207246.1 | - | 4 | 163 | |||
| BC010255.1 | - | 4 | 218 | 3709255 | ||

Ptbp2
Genomic Map