siRNA Details
| Gene Symbol: Ddx21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019553.2 | NP_062426.2 | 2 | 489 | ||
| GenBank mRNA | AF159131.3 | - | 2 | 450 | ||
| AK010281.1 | - | 2 | 369 | |||
| AK145578.1 | - | 2 | 473 | |||
| AK160095.1 | - | 2 | 463 | |||
| BC030895.1 | - | 1 | 32 | |||
| BC043655.1 | - | 2 | 461 | 5363948 | ||
| BC059237.1 | - | 2 | 449 | 6412782 | ||
| BC060220.1 | - | 2 | 441 | |||
| CF182162.1 | - | 2 | 617 | |||

Ddx21
Genomic Map