siRNA Details
| Gene Symbol: Cited4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019563.2 | NP_062509.1 | 1 | 1180 | ||
| GenBank mRNA | AF143369.1 | - | 1 | 1148 | ||
| AK012136.1 | - | 1 | 1172 | |||
| AK154030.1 | - | 1 | 1171 | |||
| BC025116.1 | - | 2 | 852 | |||
| BC080723.1 | - | 2 | 937 | |||

Cited4
Genomic Map