siRNA Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019571.5 | NP_062517.1 | 2, 3 | 513 | ||
| GenBank mRNA | AF121344.1 | - | 2, 3 | 528 | ||
| AK013350.1 | - | 2, 3 | 478 | |||
| AK015705.1 | - | 2, 3 | 235 | |||
| AK033543.1 | - | 2, 3 | 212 | |||
| AK187703.1 | - | 1, 2 | 27 | |||
| AK210177.1 | - | 1, 2 | 47 | |||
| BC046784.1 | - | 2, 3 | 416 | 6416107 | ||
| BC058695.1 | - | 2, 3 | 484 | 6813872 | ||

Tspan5
Genomic Map