siRNA Details
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019634.2 | NP_062608.2 | 4 | 401 | ||
GenBank mRNA | AF052492.1 | - | 4 | 349 | ||
AK004776.1 | - | 4 | 361 | |||
AK147306.1 | - | 4 | 363 | |||
AK147404.1 | - | 4 | 365 | |||
BC026516.1 | - | 3 | 200 | 4911317 | ||
BC050153.1 | - | 4 | 349 | |||
BC058603.1 | - | 4 | 357 | 6529011 | ||
BY238972.1 | - | 4 | 401 | |||
D26483.1 | - | 4 | 349 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108815, BC158792, BC079470, EF688600.
