siRNA Details
| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002241.4 | NP_002232.2 | 2 | 4281 | ||
| GenBank mRNA | BC034036.1 | - | 2 | 4177 | ||
| BC037840.2 | - | 1 | 1291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNJ10| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002241.4 | NP_002232.2 | 2 | 4281 | ||
| GenBank mRNA | BC034036.1 | - | 2 | 4177 | ||
| BC037840.2 | - | 1 | 1291 | |||