siRNA Details
| Gene Symbol: Pdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019501.3 | NP_062374.2 | 4 | 496 | ||
| GenBank mRNA | AB210841.1 | - | 4 | 416 | ||
| AF118855.1 | - | 2 | 191 | |||
| AK012307.1 | - | 4 | 424 | |||
| BC026820.1 | - | 4 | 398 | 4039464 | ||
| BC107273.1 | - | 4 | 311 | |||
| BC107274.1 | - | 4 | 315 | |||

Pdss1
Genomic Map