siRNA Details
| Gene Symbol: Trpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019510.2 | NP_062383.2 | 3 | 1161 | ||
| GenBank mRNA | AF190645.1 | - | 2 | 751 | ||
| AK032317.1 | - | 3 | 1162 | |||
| AK080619.1 | - | 3 | 1149 | |||
| BC108988.2 | - | 2 | 750 | |||
| BC108989.2 | - | 2 | 751 | 40047766 | ||
| FJ207475.1 | - | 2 | 751 | |||
| FJ207476.1 | - | 2 | 751 | |||

Trpc3
Genomic Map