siRNA Details
| Gene Symbol: Bace2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019517.4 | NP_062390.3 | 1, 2 | 382 | ||
| GenBank mRNA | AF216310.1 | - | 1, 2 | 333 | ||
| AK052309.1 | - | 1, 2 | 369 | |||
| AK078770.1 | - | 1, 2 | 349 | |||
| AK165036.1 | - | 1, 2 | 318 | |||
| AK186833.1 | - | 1 | 16 | |||
| BB857972.1 | - | 1, 2 | 382 | |||
| BC120773.1 | - | 1, 2 | 348 | 40129696 | ||
| BC131947.1 | - | 1, 2 | 348 | 40130224 | ||

Bace2
Genomic Map