siRNA Details
| Gene Symbol: Msgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019544.1 | NP_062417.1 | 1 | 336 | ||
| GenBank mRNA | AF261105.1 | - | 1 | 336 | ||
| BC116996.1 | - | 1 | 360 | 40126315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msgn1| Gene Symbol: Msgn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019544.1 | NP_062417.1 | 1 | 336 | ||
| GenBank mRNA | AF261105.1 | - | 1 | 336 | ||
| BC116996.1 | - | 1 | 360 | 40126315 | ||