siRNA Details
| Gene Symbol: Il17rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019583.3 | NP_062529.2 | 5 | 557 | ||
| GenBank mRNA | AF208108.1 | - | 5 | 470 | ||
| AF208109.1 | - | 5 | 470 | |||
| AK045627.1 | - | 5 | 558 | |||
| AK050139.1 | - | 5 | 478 | |||
| BC026546.1 | - | 5 | 457 | 5042466 | ||

Il17rb
Genomic Map