siRNA Details
| Gene Symbol: Styx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019637.3 | NP_062611.2 | 6, 7 | 635 | ||
| GenBank mRNA | AK002822.1 | - | 577 | |||
| AK030122.1 | - | 6, 7 | 635 | |||
| AK042741.1 | - | 7, 8 | 690 | |||
| AK046970.1 | - | 6, 7 | 612 | |||
| AK154136.1 | - | 6, 7 | 730 | |||
| AK166043.1 | - | 6, 7 | 618 | |||
| BC038608.1 | - | 6, 7 | 585 | 5012303 | ||
| BC096674.1 | - | 5, 6 | 545 | 5144003 | ||
| U34973.1 | - | 6, 7 | 603 | |||

Styx
Genomic Map